Would you like to log in again?
Ubiquitin protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U48 009-001-U48S
75933-398 75933-396
Ubiquitin protein-HIS Epitope, Rockland Immunochemicals
Ubiquitin control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Ubiquitin is a highly-conserved protein that is widely expressed in eukaryotes (1). Ubiquitin is required for ATP-dependent, nonlysosomal intracellular protein degradation of abnormal and normal proteins with a rapid turnover. A mutant form of ubiquitin, UBB+1,accumulates in the pathological hallmarks of tauopathies, including Alzheimer's disease, polyglutamine diseases, liver disease and muscle disease and serves as an endogenous reporter for proteasomal dysfunction in these diseases. By inhibiting the proteasome machinery, UBB+1 is pathogenic, since it promotes accumulation of cytotoxic misfolded amyloid-beta and phosphorylated-tau (2). Ubiquitin Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cell Cycle, Cellular Stress, Inflammation, Invasion/Metastasis, and Neurobiology research.
Ubiquitin Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol. Ubiquitin Protein is suitable for use in Western Blot. Expect a band approximately ~10kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty