Would you like to log in again?
UBE2W protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U46S 009-001-U46
75933-384EA 196.96 USD InStock
75933-384 75933-386
UBE2W protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
UBE2W control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2W or ubiquitin-conjugating enzyme E2W (putative) encodes a nuclear-localized ubiquitin-conjugating enzyme (E2) (1). Ubiquitin-conjugating enzyme (E2) along with ubiquitin-activating (E1) and ligating (E3) enzymes coordinates the addition of an ubiquitin moiety to existing proteins. UBE2W interacts with FANCL and regulates the monoubiquitination of Fanconi anemia protein FANCD2 and may be important in the repair of DNA damage (2). UBE2W is highly expressed in heart, brain, liver, and pancreas. UBE2W Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2W Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. UBE2W Protein is suitable for use in Western Blot. Expect a band approximately ~45kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty