Would you like to log in again?
UBE2V2 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U45S 009-001-U45
75933-380EA 196.96 USD InStock
75933-380 75933-382
UBE2V2 protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UBE2V2 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2V2 or ubiquitin-conjugating enzyme E2 variant 2 constitutes a distinct subfamily within the E2 protein family which is critical for the catalytic activity of E2s and also shares homology with ubiquitin-conjugating enzyme E2 variant 1 and yeast MMS2 gene product. UBE2V2 may be involved in the differentiation of monocytes and enterocytes (1). UBE2V2 functions in assembly of novel polyubiquitin chains for DNA repair (2). UBE2V2 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2V2 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2V2 Protein is suitable for use in Western Blot. Expect a band approximately ~18kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty