Would you like to log in again?
UBE2U protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U44S 009-001-U44
75933-376EA 353.41 USD InStock
75933-376 75933-378
UBE2U protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
UBE2U control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2U, or Ubiquitin-conjugating enzyme E2 U, belongs to the ubiquitin-conjugating enzyme family. In the global yeast two-hybrid (Y2H) screens to study the specificity of interactions between catalytic domains of the human E2s with RING-type E3s, the analysis showed the UBE2U had a high-quality and physical interactions with a few E3 ligases (1) and (2). UBE2U Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2U Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. UBE2U Protein is suitable for use in Western Blot. Expect a band approximately ~52kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty