Would you like to log in again?
UBE2R2(UBC3B) protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U43S 009-001-U43
75933-372EA 196.96 USD InStock
75933-372 75933-374
UBE2R2(UBC3B) protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UBE2R2(UBC3B) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2R2 (UBC3B) or ubiquitin-conjugating enzyme E2R 2 encodes a protein similar to the E2 ubiquitin conjugating enzyme UBC3/CDC34. CK2-dependent phosphorylation of this ubiquitin-conjugating enzyme functions by regulating beta-TrCP substrate recognition and induces its interaction with beta-TrCP thereby enhancing beta-catenin degradation. CK2-dependent phosphorylation of CDC34 and UBC3B functions by regulating BTRC substrate recognition (1). UBE2R2 complements a yeast cdc34 temperature-sensitive mutant. Deletion and site-directed mutagenesis demonstrated that CK2 phosphorylated UBE2R2 in the C-terminal domain at serine-233; mutagenesis of the corresponding serine residue at position 231 of human CDC34/UBC3 showed that this was the major CK2-phosphorylation site in this protein. UBE2R2 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2R2 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2R2 Protein is suitable for use in Western Blot. Expect a band approximately ~32kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty