Would you like to log in again?
UBE2R1 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U42 009-001-U42S
75933-370EA 307.26 USD InStock
75933-370 75933-368
UBE2R1 protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UBE2R1 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2R1 is a member of the ubiquitin-conjugating enzyme family which catalyzes the covalent attachment of ubiquitin to other proteins. UBE2R1 is a part of the large multiprotein complex which is required for ubiquitin-mediated degradation of cell cycle G1 regulators and for the initiation of DNA replication (1). CK2 can phosphorylate CDC34 and UBC3B at corresponding serine residue in the C terminus of each protein. Phosphorylated UBC3B and CDC34 binds specifically to the F-box protein beta-TRCP which results in enhanced degradation of beta-catenin, a substrate of BTRC. CK2-dependent phosphorylation of CDC34 and UBC3B functions by regulating BTRC substrate recognition (2). UBE2R1 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2R1 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2R1 Protein is suitable for use in Western Blot. Expect a band approximately ~33kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty