Would you like to log in again?
UBE2L6(UBCH8) protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U40 009-001-U40S
75933-358EA 307.26 USD InStock
75933-358 75933-356
UBE2L6(UBCH8) protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UBE2L6(UBCH8) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2L6 (UBCH8) or ubiquitin-conjugating enzyme E2L 6 is a member of the E2 ubiquitin-conjugating enzyme family which is highly similar in primary structure to the enzyme encoded by the UBE2L3 gene. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s) and the selective physical interaction between E2 and E3 enzymes forms the basis of specificity for functionally distinct E2-E3 combinations (1). UBE2L6 and the ubiquitin-ligase SIAH1 physically interact with and target FLT3-ITD for proteasomal degradation (2). UBE2L6 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2L6 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2L6 Protein is suitable for use in Western Blot. Expect a band approximately ~17kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty