Would you like to log in again?
UBE2J1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U38S 009-001-U38
75933-348EA 353.41 USD InStock
75933-348 75933-350
UBE2J1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
UBE2J1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2J1 is a member of the E2 ubiquitin-conjugating enzyme family which is located in the membrane of the endoplasmic reticulum (ER) and may contribute to quality control ER-associated degradation by the ubiquitin-proteasome system. UBE2J1 play an important role in the homeostatic regulation of MHC class I assembly and expression (1). HRD1 and UBE2J1 associate in a complex together with non-β2m bound MHC class I heavy chains, Derlin 1, and p97 and discriminate misfolded MHC class I from conformational MHC I-β2m-peptide heterotrimers. UBE2J1 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2J1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. UBE2J1 Protein is suitable for use in Western Blot. Expect a band approximately ~70kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty