Would you like to log in again?
UBE2G1 (UBC7) protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U35 009-001-U35S
75933-338EA 307.26 USD InStock
75933-338 75933-336
UBE2G1 (UBC7) protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UBE2G1 (UBC7) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2G1 (UBC7) or ubiquitin-conjugating enzyme E2G 1 is a member of the E2 ubiquitin-conjugating enzyme family and catalyzes the covalent attachment of ubiquitin to other proteins which are involved in degradation of muscle-specific proteins. UBC7 is highly expressed in skeletal muscle (1). UBC7 is involved in the selective ubiquitylation of different substrates by the same E3 complex (2). UBE2G1 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2G1 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2G1 Protein is suitable for use in Western Blot. Expect a band approximately ~21kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty