Would you like to log in again?
UBE2F protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U34S 009-001-U34
75933-332EA 196.96 USD InStock
75933-332 75933-334
UBE2F protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UBE2F control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2F or ubiquitin-conjugating enzyme E2F is part of the ubiquitin network for protein degradation and is involved in protein neddylation. UBE2F has been shown to function as a NEDD8-conjugating enzyme in vitro and in vivo (1). Covalent attachment of the ubiquitin-like protein NEDD8 to a conserved C-terminal domain (ctd) lysine stimulates Cullin Ring ligase (CRL) ubiquitination activity and prevents binding of the inhibitor CAND1 (2). UBE2F Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2F Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2F Protein is suitable for use in Western Blot. Expect a band approximately ~24kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty