Would you like to log in again?
UBE2E3(UBCH9) protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U33S 009-001-U33
75933-328EA 196.96 USD InStock
75933-328 75933-330
UBE2E3(UBCH9) protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UBE2E3(UBCH9) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBE2E3 also known as UBCH9 or ubiquitin-conjugating enzyme E2E 3 is a member of the E2 ubiquitin-conjugating enzyme family. Ubiquitination, the covalent attachment of the protein ubiquitin (Ub) to other cellular proteins is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation that involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s(1). UBE2E3 is highly expressed in skeletal muscle (2). UBE2E3 is essential for the proliferation of RPE-1 cells and is down regulated during RPE layer maturation in the developing mouse eye. These findings indicate that UBE2E3 is a major enzyme in modulating the balance between RPE cell proliferation and differentiation. UBE2E3 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBE2E3 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UBE2E3 Protein is suitable for use in Western Blot. Expect a band approximately ~25kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty