Would you like to log in again?
UBA2(SAE2) protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U25S 009-001-U25
75935-478EA 353.41 USD InStock
75935-478 75935-480
UBA2(SAE2) protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
UBA2(SAE2) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBA2 (SAE2) or ubiquitin-like modifier activating enzyme 2 forms a heterodimer that functions as a SUMO-activating enzyme for the sumoylation of proteins. The posttranslational modification of proteins by the addition of the small protein SUMO, or sumoylation, regulates protein structure and intracellular localization (1). UBA2 functions in SUMO1 activation in a manner analogous to the single E1 ubiquitin-activating enzymes (2). The inactivation of UBA2 may be a therapeutic strategy in MYC-driven cancers. UBA2 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBA2 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. UBA2 Protein is suitable for use in Western Blot. Expect a band approximately ~115kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty