Would you like to log in again?
TRDMT1 (DNMT2) protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U19 009-001-U19S
75933-090EA 665.18 USD InStock
75933-090 75933-088
TRDMT1 (DNMT2) protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
TRDMT1 (DNMT2) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TRDMT1 is responsible for the methylation of aspartic acid transfer RNA, specifically at the cytosine-38 residue in the anticodon loop. TRDMT1 also possesses residual DNA-(cytosine-C5) methyltransferase activity. While similar in sequence and structure to DNA cytosine methyltransferases, TRDMT1 is distinct and highly conserved in its function among various species. The indirect sequence recognition is also a feature of eukaryotic DNA methyltransferases, which may have arisen from a DNMT2-like RNA methyltransferase (1). DNMT2 is involved in retrotransposon silencing and telomere integrity in somatic cells (2). TRDMT1 Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, and Neurobiology research.
TRDMT1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TRDMT1 Protein is suitable for use in Western Blot. Expect a band approximately ~67kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty