Would you like to log in again?
TRAF2 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U18 009-001-U18S
75933-032 75933-030
TRAF2 protein-HIS Epitope, Rockland Immunochemicals
TRAF2 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TRAF2 (also known as TNF receptor-associated factor 2) belongs to the TNF receptor associated factor (TRAF) protein family that mediates signal transduction from members of the TNF receptor superfamily (1). TRAF2 is involved in TNF-alpha-mediated activation of MAPK8/JNK and NF-kB pathway. TRAF2 interacts with TNF receptors and forms a heterodimeric complex with TRAF1. The TRAF1-TRAF2 complex interacts with IAPs to mediate the anti-apoptotic signals from TNF receptors. TRAF2 also interacts with TRADD, a TNF receptor associated apoptotic signal transducer, to ensure the recruitment of IAPs for the direct inhibition of caspase activation (2). TRAF2 Protein is ideal for investigators involved in Signaling Proteins, Adaptor Proteins, Apoptosis/Autophagy, Cancer, Inflammation, JNK/SAPK Pathway, and NfkB Pathway research.
TRAF2 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. TRAF2 Protein is suitable for use in Western Blot. Expect a band approximately ~59kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty