Would you like to log in again?
TPX2 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U17 009-001-U17S
75935-466EA 472.72 USD InStock
75935-466 75935-464
TPX2 protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
TPX2 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TPX2 is a nuclear proliferation-associated protein whose expression is restricted to cell cycle phases S, G2, and M (1). It mainly expressed in the lung carcinoma cell lines and also found in adult placenta, skeletal muscle, thymus, testis, and small intestine and in fetal brain, liver, and kidney.TPX2 is required for targeting STK6 to the spindle apparatus, and that STK6 may regulate the function of TPX2 during spindle assembly. TPX2 functions mainly in mitotic spindle assembly and likely contributes to its role in chromosome stability control and tumor suppression (2). TPX2 Protein is ideal for investigators involved in Signaling Proteins, Microtubule/Actin Associated Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, Invasion/Metastasis, and Neurobiology research.
TPX2 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. TX2 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~110kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty