TP53RK protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U16S 009-001-U16
75933-026EA 364.21 USD
75933-026 75935-460
TP53RK protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
TP53RK control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TP53RK or TP53 regulating kinase and also called PRPK (p53-related protein kinase) is the human homologue of yeast Bud32. TP53RK belongs to a small subfamily of atypical protein kinases. TP53RK is expressed in interleukin-2-activated cytotoxic T-cells, epithelial tumor cell lines, and the testes and lower expression of TP53RK is found in heart, kidney, and spleen (1). PRPK interacts with CGI-121 and co-precipitation of p53 with PRPK is inhibited by adding recombinant CGI-121 in vitro. CGI-121 may act as an inhibitor of the PRPK-p53 interaction (2). TP53RK is activated by another kinase, Akt/PKB, which phosphorylates TP53RK at Ser250. TP53RK Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, Cellular Stress, Inflammation, JNK/SAPK Pathway, Metabolic Disorder, Neurobiology, and p38 Pathway research.
TP53RK Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TP53RK Protein is suitable for use in Western Blot. Expect a band approximately ~55kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.