Would you like to log in again?
TET3 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U15 009-001-U15S
75932-838 75932-836
TET3 protein-GST fusion, Rockland Immunochemicals
TET3 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TET3 or Tet methylcytosine deoxygenase 3 is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. TET3 which is a member of the ten-eleven translocation (TET) gene family is involved in myelopoiesis, and defects in TET3 have been associated with several myeloproliferative disorders. TET plays an important role in embryonic stem cell maintenance and inner cell mass cell specification (1). TET3 play an important role in the DNA methylation process (2). TET3 Protein is ideal for investigators involved in Signaling Proteins, Methylcytosine Dioxygenase Proteins, Cancer, and Cell Cycle research.
TET3 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TET3 Protein is suitable for use in Western Blot. Expect a band approximately ~140kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty