Would you like to log in again?
TET2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U14 009-001-U14S
75932-834EA 785.6 USD InStock
75932-834 75932-832
TET2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
TET2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TET2 is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine which is involved in myelopoiesis. Defects in TET2 gene have been associated with several myeloproliferative disorders. TET2 play an important role in embryonic stem cell maintenance and inner cell mass cell specification. TET proteins have potential roles in epigenetic regulation through modification of 5mC to 5hmC (1). TET2 enzymatic activity is involved in myeloid tumorigenesis and this could be valuable as a diagnostic and prognostic tool for targeted therapies and to assess responses to anticancer drugs (2). TET2 Protein is ideal for investigators involved in Signaling Proteins, Methylcytosine Dioxygenase Proteins, Cancer, and Cell Cycle research.
TET2 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TET2 Protein is suitable for use in Western Blot. Expect a band approximately ~124kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty