Would you like to log in again?
TESK1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U12S 009-001-U12
75932-826EA 353.41 USD InStock
75932-826 75935-436
TESK1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
TESK1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TESK1 or testis-specific kinase 1 is a serine/threonine protein kinase that contains an N-terminal protein kinase domain and a C-terminal proline-rich domain which is most closely related to those of the LIM motif-containing protein kinases (LIMKs). TESK1 protein can phosphorylate myelin basic protein and histone in vitro and plays an important role at and after the meiotic phase of spermatogenesis. TESK1 is mainly expressed in testicular germ cells (1). TESK1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, and Ser/Thr Kinases research.
TESK1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TESK1 Protein is suitable for use in Western Blot. Expect a band approximately ~106kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty