MAPT
Supplier: Rockland Immunochemicals
009-001-T75 009-001-T75S
75932-626EA 513.11 USD
75932-626 75932-624
MAPT
Proteins and Peptides
Tau-383 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Tau-383 or Tau-D is a member of the Tau family of proteins which function to stabilize the microtubules by binding to them. Tau proteins are subject to phosphorylation and this phenomenon regulates the association of the Tau protein with the microtubules (1). Deposits of Alzheimer's disease AD-associated proteins, such as hyperphosphorylated Tau, as well as other shared misfolded proteins, such as, β-amyloid precursor protein (βAPP), ubiquitin, and various chaperones and protein kinases are thought to play a pathologic role in the cognitive decline and muscular failure. Malfunctioning of Tau proteins is associated with microtubules disintegration and collapsing of the neuronal transport system (2). Tau-383 Protein is ideal for investigators involved in Signaling Proteins, Tau Proteins, Invasion/Metastasis, Neurobiology, and p38 Pathway research.
Tau-383 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. Tau-383 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~52kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.