Would you like to log in again?
TAF1 (C-term) protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T70 009-001-T70S
75932-558EA 665.18 USD InStock
75932-558 75932-556
TAF1 (C-term) protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
TAF1 (C-term) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TAF1 is a DNA-binding protein complex required for RNA polymerase II-mediated transcription of many, if not all, protein-encoding genes in eukaryotic cells. TAF1 plays a key role in the initiation process, since it binds to the TATA element to form a complex that nucleates the assembly of the other components into a preinitiation complex and that may be stable through multiple rounds of transcription. TAF1 may be targeted to specific chromatin-bound promoters and may play a key role in chromatin recognition (1). TAF1 also serves to link the control of transcription to the cell cycle (2). TAF1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cell Cycle, and Invasion/Metastasis research.
TAF1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TAF1 Protein is suitable for use in Western Blot. Expect a band approximately ~155kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty