Would you like to log in again?
STK9 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T59 009-001-T59S
75932-310EA 665.18 USD InStock
75932-310 75932-308
STK9 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
STK9 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
STK9 or CDKL5 is a member of Ser/Thr protein kinase family which has protein kinase activity and is associated with X-linked infantile spasm syndrome (ISSX), also known as X-linked West syndrome, and Rett syndrome (RTT). STK9 can control nuclear speckle morphology by regulating the phosphorylation state of splicing regulatory proteins, and may be involved indirectly in pre-mRNA processing, by controlling splicing factor dynamics (1). The mutations in the STK9 are a major cause of infantile spasms and early epileptic seizures in female patients, and of a later intractable seizure disorder (2). STK9 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cardiovascular Disease, ERK/MAPK Pathway, Neurobiology, PKA/PKC Pathway, and Ser/Thr Kinases research.
STK9 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. STK9 Protein is suitable for use in Western Blot. Expect a band approximately ~84kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty