STAT6
Supplier: Rockland Immunochemicals
009-001-T57S 009-001-T57
75932-224EA 713.69 USD
75932-224 75932-222
STAT6
Proteins and Peptides
STAT6 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
The STAT6 protein is a member of the STAT family of transcription factors that are activated by cytokines and phosphorylated by the receptor associated kinases. STATs form homo- or hetero-dimers that translocate to the cell nucleus where they act as transcription activators (1). STAT6 plays a central role in exerting IL4 mediated biological responses. STAT6 induces the expression of BCL2L1/BCL-XL, which is responsible for the anti-apoptotic activity of IL4. STAT6 mRNA has been detected in various tissues including peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response (2). STAT6 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, Inflammation, and JAK/STAT Pathway research.
STAT6 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. STAT6 Protein is suitable for use in Western Blot. Expect a band approximately ~138kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.