STAT3 protein-HIS Epitope, Rockland Immunochemicals
009-001-T54 009-001-T54S
75932-202EA 658.59 USD
75932-202 75932-200
STAT3 protein-HIS Epitope, Rockland Immunochemicals
STAT3 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
STAT3 is a member of the signal transducers and activators of transcription (STAT) family of proteins that carry out a dual function: signal transduction and activation of transcription. STAT3 is widely expressed and becomes activated through phosphorylation on tyrosine as a DNA binding protein in response to a variety of stimuli such as EGF, IL-6, PDGF, IL-2 and G-CSF (1). This phosphoprotein forms homodimers as well as heterodimers with other members of the STAT family and translocate to the nucleus in order to modulate the transcription of various genes (2). STAT3 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, Inflammation, and JAK/STAT Pathway research.
STAT3 Protein is stored in 50mM NaPhosphate pH7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol. STAT3 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~95kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.