Would you like to log in again?
STAT3 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T53S 009-001-T53
75932-196EA 353.41 USD InStock
75932-196 75932-198
STAT3 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
STAT3 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
STAT3 is a member of the signal transducers and activators of transcription (STAT) family of proteins that carry out a dual function: signal transduction and activation of transcription. STAT3 is widely expressed and becomes activated through phosphorylation on tyrosine as a DNA binding protein in response to a variety of stimuli such as EGF, IL-6, PDGF, IL-2 and G-CSF (1). This phosphoprotein forms homodimers as well as heterodimers with other members of the STAT family and translocate to the nucleus in order to modulate the transcription of various genes (2). STAT3 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, Inflammation, and JAK/STAT Pathway research.
STAT3 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. STAT3 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~120kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty