Would you like to log in again?
SMYD3 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T44 009-001-T44S
75932-064 75932-062
SMYD3 protein-GST fusion, Rockland Immunochemicals
SMYD3 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SMYD3 (SET and MYND domain containing 3) is a histone methyltransferase that plays a role in transcriptional regulation as a member of an RNA polymerase complex. The introduction of SMYD3 enhanced cell growth and the histone methyltransferase activity of SMYD3 involved in the proliferation of cancer cells (1). A variable number of tandems repeat polymorphism in an E2F-1 binding element in the 5-prime flanking region of SMYD3 is a risk factor for human cancers (2). SMYD3 Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Cancer, Cell Cycle, and Neurobiology research.
SMYD3 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SMYD3 Protein is suitable for use in Western Blot. Expect a band approximately ~71kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty