Would you like to log in again?
SMYD2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T43 009-001-T43S
75932-060EA 665.18 USD InStock
75932-060 75932-058
SMYD2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
SMYD2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SMYD2 (also known as SET and MYND domain-containing protein 2) is a member of the SMYD family of protein methyltransferases. SMYD2 is also known as lysine methyltransferase protein 3C (KMT3C). SMYD2 functions to repress transcription by interacting with the Sin3A histone deacetylase complex and performs methylation of Lys36 in histone H3 (1). The exogenous expression of SMYD2 in mouse fibroblasts decreases cell growth. SMYD2 also function as a putative oncogene by methylating p53 and repressing its tumor suppressive function (2). SMYD2 Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, and Neurobiology research.
SMYD2 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SMYD2 Protein is suitable for use in Western Blot. Expect a band approximately ~70kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty