Would you like to log in again?
SMAD5 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T41 009-001-T41S
75932-046EA 665.18 USD InStock
75932-046 75932-044
SMAD5 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
SMAD5 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SMAD5 is a member of the SMAD family and mediates signaling by the transforming growth factor-beta (TGFβ) superfamily and related ligands (1). SMAD5 plays a critical role in the signaling pathway by which TGFβ inhibits the proliferation of human hematopoietic progenitor cells. SMAD5 is up-regulated in gastric epithelial cells during the infection of the pathogen Helicobacter pylori and it mediates apoptosis of gastric epithelial cells induced by H. pylori infection (2). In mature human B cells, bone morphogenetic protein 6 (BMP-6) inhibits cell growth and rapidly induces phosphorylation of SMADs 5 and 8. SMAD5 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, AKT/PKB Pathway, Angiogenesis, Cancer, Cell Cycle, Cellular Stress, ERK/MAPK Pathway, Inflammation, JAK/STAT Pathway, JNK/SAPK Pathway, NfkB Pathway, and WNT Signaling research.
SMAD5 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SMAD5 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~82kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty