SMAD4
Supplier: Rockland Immunochemicals
009-001-T40S 009-001-T40
75932-042EA 713.69 USD
75932-042 75932-040
SMAD4
Proteins and Peptides
SMAD4 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SMAD4 is a member of the SMAD family and mediates signaling by the transforming growth factor-beta (TGFβ) superfamily and related ligands (1). TGFβ stimulation leads to phosphorylation and activation of SMAD1, SMAD2 and SMAD3, which form complexes with SMAD4 that accumulate in the nucleus and regulate transcription of target genes. SMAD signaling is negatively regulated by inhibitory SMADs and ubiquitin-mediated processes and proteasomal degradation of SMADs depend on the direct interaction of specific E3 ligases with SMADs. SMAD4 is targeted for degradation by multiple ubiquitin ligases that can simultaneously act on R-SMADs and signaling receptors. Such mechanisms of down-regulation of TGFβ signaling via degradation of SMADs may be critical for proper physiological response to this pathway (2). SMAD4 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, AKT/PKB Pathway, Angiogenesis, Cancer, Cell Cycle, Cellular Stress, ERK/MAPK Pathway, Inflammation, JAK/STAT Pathway, JNK/SAPK Pathway, NfkB Pathway, and WNT Signaling research.
SMAD4 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SMAD4 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~95kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.