Would you like to log in again?
SIRT5 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T35S 009-001-T35
75931-932 75931-934
SIRT5 protein-GST fusion, Rockland Immunochemicals
SIRT5 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SIRT5 is a member of the sirtuin family of proteins which are homologs to the yeast Sir2 protein. Sirtuin family contain a sirtuin core domain and are grouped into four classes with SIRT5 being a member of class III. SIRT5 consists of eight exons and is found in two isoforms which encode a 310 aa and a 299 aa protein, respectively. Human SIRT5 is most predominantly expressed in heart muscle cells and in lymphoblasts. Fluorescence in situ hybridization analysis localized the human SIRT5 gene to chromosome 6p23. SIRT5 can deacetylate cytochrome c, a protein of the mitochondrial intermembrane space with a central function in oxidative metabolism as well as apoptosis initiation (1). SIRT5 Protein is ideal for investigators involved in Signaling Proteins, Deacetylase/Demethylase Proteins, Cancer, Cell Cycle, and Inflammation research.
SIRT5 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SIRT5 Protein is suitable for use in Western Blot. Expect a band approximately ~60kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty