SIRT3 control protein-GST fusion
- Control Protein
- PCA Assays
- ImmunoBlot
SIRT3 is a member of the sirtuin family of proteins which are homologs to the yeast Sir2 protein. Sirtuin family contain a sirtuin core domain and are grouped into four classes with SIRT3 being a member of class I. SIRT3 contains an N-terminal mitochondrial targeting signal and a central catalytic domain and localizes in the nuclei and mitochondria (1). SIRT3 has NAD(+)-dependent histone deacetylase activity with specificity for lys16 of H4 and, to a lesser extent, lys9 of H3. SIRT3 can repress transcription of a target gene in vivo when recruited to its promoter. Full-length SIRT3 is transported from the nucleus to the mitochondria during cell stress conditions (2). SIRT3 Protein is ideal for investigators involved in Signaling Proteins, Deacetylase/Demethylase Proteins, Cancer, Cell Cycle, and Inflammation research.
SIRT3 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SIRT3 Protein is suitable for use in Western Blot. Expect a band approximately ~66kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.