SCYL1
Supplier: Rockland Immunochemicals
009-001-T26 009-001-T26S
75931-756EA 713.69 USD
75931-756 75931-754
SCYL1
Proteins and Peptides
SCYL1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SCYL1 is a transcriptional regulator belonging to the SCY1-like family of kinase-like proteins. SCYL1 has a divergent N-terminal kinase domain that is thought to be catalytically inactive, and can bind specific DNA sequences through its C-terminal domain. SCYL1 activates transcription of the telomerase reverse transcriptase and DNA polymerase beta genes. SCYL1 forms multimers following transfection into COS-7 cells (1). A SCYL1 binding protein has been identified that co-localized with SCYL1 in cytoplasm and shows ubiquitous expression (2). SCYL1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Inflammation, Invasion/Metastasis, and Neurobiology research.
SCYL1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SCYL1 Protein is suitable for use in Western Blot. Expect a band approximately ~85kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.