Would you like to log in again?
RYK protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T25S 009-001-T25
75931-710EA 353.41 USD InStock
75931-710 75931-712
RYK protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
RYK control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
RYK or receptor-like tyrosine kinase is an atypical member of the family of growth factor receptor protein tyrosine kinases. RYK differs from other receptor tyrosine kinases at a number of conserved residues in the activation and nucleotide binding domains (1). RYK has a leucine-rich extracellular domain with a WIF-type WNT binding region, a single transmembrane domain, and an intracellular tyrosine kinase domain. RYK is involved in stimulating WNT signaling pathways such as the regulation of axon path finding (2). RYK Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Neurobiology, Receptor Tyrosine Kinases, and WNT Signaling research..
RYK Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. RIYK Protein is suitable for use in Western Blot. Expect a band approximately ~66kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty