Would you like to log in again?
RhoD protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T23 009-001-T23S
75931-594 75931-592
RhoD protein-HIS Epitope, Rockland Immunochemicals
RhoD control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
RhoD is a member of the Rho gene family that interacts with protein kinases and serves as a target of activated GTPase. Rho binds GTP and plays a critical role in muscle differentiation by regulating the expression of myogenin and MEF2 genes. Furthermore, Rho is involved in endosome dynamics and reorganization of the actin cytoskeleton and plays a role in coordinating membrane transport (1). mDia and ROCK are downstream effectors of Rho mediating Rho action on the actin cytoskeleton; mDia produces actin filaments by nucleation and polymerization and ROCK activate myosin to cross-link them for induction of actomyosin bundles and contractility (2). RhoD Protein is ideal for investigators involved in Signaling Proteins, G-Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, ERK/MAPK Pathway, Invasion/Metastasis, and Neurobiology research.
RhoD Protein is stored in 50mM MOPS, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. RhoD Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~22kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty