Would you like to log in again?
RhoA protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T21S 009-001-T21
75931-584 75931-586
RhoA protein-HIS Epitope, Rockland Immunochemicals
RhoA control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
RhoA is a member of the Rho gene family that interacts with protein kinases and serves as a target of activated GTPase. RhoA play an important role in the initial events of cell protrusion, whereas Rac1 and Cdc42 activate pathways implicated in reinforcement and stabilization of newly expanded protrusions. Rho mediates corneal epithelial migration in response to external stimuli by regulating the organization of the actin cytoskeleton (1). The inhibition of RhoA signaling is required for both vaccinia morphogenesis and virus-induced cell motility. Vaccinia inhibits RhoA signaling using the viral protein F11 and this can enhance the spread of infection (2). RhoA Protein is ideal for investigators involved in Signaling Proteins, G-Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, ERK/MAPK Pathway, Invasion/Metastasis, and Neurobiology research.
RhoA Protein is stored in 50mM MOPS, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. RhoA Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~23kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty