Would you like to log in again?
RGS1 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T19 009-001-T19S
75931-562 75931-560
RGS1 protein-HIS Epitope, Rockland Immunochemicals
RGS1 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
RGS1 is a member of the regulator of G-protein signaling family and attenuates the signaling activity of G-proteins. RGS1 binds to activated GTP-bound G alpha subunit and acts as a GTPase activating protein (GAP) thereby increasing the rate of conversion of GTP to GDP and terminating the signal (1). RGS1 is extensively up-regulated in renal cell carcinoma (RCC) tissues and melanoma. In melanoma, RGS1 expression is significantly correlated with increased tumor thickness, mitotic rate and presence of vascular involvement. Furthermore, there is significant association between increasing RGS1 expression and reduced relapse-free survival as well as disease-specific survival (DSS) survival (2). RGS1 Protein is ideal for investigators involved in Signaling Proteins, G-Proteins, AKT/PKB Pathway, Cancer, Cell Cycle, ERK/MAPK Pathway, Inflammation, JNK/SAPK Pathway, p38 Pathway, and PKA/PKC Pathway research.
RGS1 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. RGS1 Protein is suitable for use in Western Blot. Expect a band approximately ~26kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty