Would you like to log in again?
Rb protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T17 009-001-T17S
75935-210EA 665.18 USD InStock
75935-210 75935-206
Rb protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
Rb control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Rb is the retinoblastoma gene product and was the first tumor suppressor cloned. Rb is a negative regulator of the cell cycle through its ability to bind the transcription factor E2F and repress transcription of genes required for S phase (1). Rb can bind to MDM2 and this overcomes both the antiapoptotic function of MDM2 and the MDM2-dependent degradation of p53. Rb associates with the Polycomb group (PcG) proteins to form a repressor complex that blocks entry of cells into mitosis (2). Rb colocalizes with nuclear PcG complexes and is important for association of PcG complexes with nuclear targets. Rb Protein is ideal for investigators involved in Signaling Reagents, Protein Substrates, Cancer, Cell Cycle, and Neurobiology research.
Rb Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. Rb Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~62kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty