RAC1
Supplier: Rockland Immunochemicals
009-001-T12S 009-001-T12
75934-138EA 713.69 USD
75934-138 75934-136
RAC1
Proteins and Peptides
RAC1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
RAC1 is a member of the Rho family and is a GTPase that is part of the small GTP-binding protein superfamily. RAC1 is involved in a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases (1). Role of RAC1 in colorectal cancer has been reported after analysis of the protein expression level and activities of this protein in matched sets of tumor and non-tumor tissues. Overexpression of RAC1 leads to increased tumor growth in xenografts of human colorectal tumor cells (2). RAC1 is also involved in the regulation of critical cellular functions including organization of actin cytoskeleton, transcription control and cell cycle. RAC1 Protein is ideal for investigators involved in Signaling Proteins, G-Proteins, Cancer, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, and Neurobiology research.
RAC1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. RAC1 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~47kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.