Would you like to log in again?
PTPRM (PTP-m) protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T11 009-001-T11S
75931-392EA 665.18 USD InStock
75931-392 75931-390
PTPRM (PTP-m) protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PTPRM (PTP-m) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PTPRM is a member of the protein tyrosine phosphatase family and can participate in a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation (1). PTPRM has been shown to mediate cell-cell aggregation through the interaction with another molecule of PTPRM on an adjacent cell. PTPRM can interact with scaffolding protein RACK1/GNB2L1 and this interaction may be necessary for downstream signaling in response to cell-cell adhesion (2). PTPRM has been shown to be expressed in human pulmonary vascular endothelia where it directly binds to VE-cadherin and regulates both the tyrosine phosphorylation state of VE-cadherin and barrier integrity. PTPRM Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cardiovascular Disease, Neurobiology, and Phosphatases research.
PTPRM Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PTPRM Protein is suitable for use in Western Blot. Expect a band approximately ~110kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty