Would you like to log in again?
PTK9 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T10 009-001-T10S
75931-382EA 665.18 USD InStock
75931-382 75931-380
PTK9 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PTK9 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PTK9 is an actin monomer-binding protein conserved from yeast to mammals which has been localized to cortical G-actin-rich structures that may be regulated by the small GTPase RAC1. PTK9 has tyrosine kinase activity similar to that of recombinant FGFR2, with optimal activity over pH 6.5 to 7.4 and a preference for manganese over magnesium as a divalent cofactor (1). PTK9 plays a central role in regulation of actin dynamics and cell motility in lymphoma cell homeostasis in vivo and represent as a bona fide lymphoma drug target (2). PTK9 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, and Neurobiology research.
PTEK9 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PTK9 Protein is suitable for use in Western Blot. Expect a band approximately ~64kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty