Would you like to log in again?
PTK7 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T09 009-001-T09S
75931-378EA 665.18 USD InStock
75931-378 75931-376
PTK7 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PTK7 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PTK7 or pseudo protein tyrosine kinase 7 is a member of the receptor protein tyrosine kinase family of proteins that transduce extracellular signals across the cell membrane. PTK7 lacks detectable catalytic tyrosine kinase activity but is involved in the WNT signaling pathway and plays a role in multiple cellular processes including polarity and adhesion (1). PTK7 interacts with β-catenin and functions upstream from glycogen synthase kinase 3. PTK7 has been isolated from colon carcinoma and was designated colon carcinoma kinase-4 (2). In conjunction with other T cell markers, PTK7 has utility as a biomarker for detecting minimal residual disease of T-ALL in the bone marrow. PTK7 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, and Neurobiology research.
PTEK7 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PTK7 Protein is suitable for use in Western Blot. Expect a band approximately ~66kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty