PP2A beta protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T01 009-001-T01S
75931-122EA 713.69 USD
75931-122 75931-120
PP2A beta protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PP2A beta control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PP2A is one of the four major Ser/Thr phosphatases which are implicated in the negative control of cell growth and division and encodes a specific phosphotyrosyl phosphatase activator of the dimeric form of protein phosphatase 2A (1). PP2A holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B which is encoded by a diverse set of genes. The interaction was independent of ATP or magnesium ion and the inclusion of PTPA altered the substrate specificity of PP2A, with enhanced phosphotyrosine phosphatase activity and decreased phosphoserine phosphatase activity (2). PP2Aβ Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cardiovascular Disease, Cell Cycle, ERK/MAPK Pathway, Metabolic Disorder, Phosphatases, Ser/Thr Kinases, and WNT Signaling research.
PP2Aβ Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PP2Aβ Protein is suitable for use in Western Blot. Expect a band approximately ~59kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.