Would you like to log in again?
POR protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T00S 009-001-T00
75934-118 75931-112
POR protein-GST fusion, Rockland Immunochemicals
POR control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
POR is a flavoprotein that donates electrons to all microsomal P450 enzymes, including the steroidogenic enzymes P450c17, P450c21, and CYP51A1. POR encodes an endoplasmic reticulum membrane oxidoreductase with an FAD-binding domain and a flavodoxin-like domain which binds two cofactors, FAD and FMN, which allow it to donate electrons directly from NADPH to all microsomal P450 enzymes (1). POR has been linked with various diseases, including apparent combined P450C17 and P450C21 deficiency, amenorrhea and disordered steroidogenesis, congenital adrenal hyperplasia and Antley-Bixler syndrome (2). POR Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Apoptosis/Autophagy, Cancer, and Metabolic Disorder research.
POR Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. POR Protein is suitable for use in Western Blot. Expect a band approximately ~102kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty