Would you like to log in again?
PIP4K2B protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S98 009-001-S98S
75931-020 75934-986
PIP4K2B protein-GST fusion, Rockland Immunochemicals
PIP4K2B control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PIP4K2B (phosphatidylinositol-5-phosphate 4-kinase, type II, beta) catalyzes the phosphorylation of phospha-tidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5, 4-bisphosphate. PIP4K2B is a member of the phosphatidylinositol-5-phosphate 4-kinase family. PIP5K2B interacts specifically with the juxtamembrane region of the p55 TNF receptor and that PIP5K2B activity is increased in mammalian cells by treatment with TNF-alpha (1). PIP5P4KB plays a role in determining insulin sensitivity and adiposity (2). PIP4K2B Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Invasion/Metastasis, Lipid Kinases, Metabolic Disorder, Neurobiology, NfkB Pathway, and WNT Signaling research.
PIP4K2B Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PIP4K2B Protein is suitable for use in Western Blot. Expect a band approximately ~73kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty