Would you like to log in again?
PIK3C2B protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S97S 009-001-S97
75930-990EA 353.41 USD InStock
75930-990 75930-992
PIK3C2B protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PIK3C2B control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PIK3C2B or phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 beta belongs to the phosphoinositide 3-kinase (PI3K) family. PIK3C2B plays a role in signaling pathways involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. PIK3C2B is highly expressed in thymus and placenta and it may function in a calcium-independent manner (1). In cell signaling, PIK3C2B stabilizes nucleotide-free Ras and the interaction of Ras and PI3KC2 beta mutually inhibits one another activity (2). PIK3C2B Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Invasion/Metastasis, Lipid Kinases, Metabolic Disorder, Neurobiology, NfkB Pathway, and WNT Signaling research.
PIK3C2B Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PIK3C2B Protein is suitable for use in Western Blot. Expect a band approximately ~98kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty