Would you like to log in again?
PI3K (p85 beta) protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S95S 009-001-S95
75930-922 75930-924
PI3K (p85 beta) protein-HIS Epitope, Rockland Immunochemicals
PI3K (p85 beta) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Phosphatidylinositol 3-kinase (PI3K) is a lipid kinase that phosphorylates the inositol ring of phosphatidylinositol and related lipid products at the 3-prime position. PI3K p85 subunit which is the regulatory subunit of PI3K and consists of 2 closely related proteins, p85-alpha and p85-beta (1). The activation of PI3K signaling in influenza A virus-infected cells is important for efficient virus replication (2). PI3K signaling in T cells can lead to organ-specific autoimmunity and the class IA PI3K-deficient mice manifest the cardinal features of human primary Sjogren’s syndrome-like disease (3). PI3k (p85 beta) Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Invasion/Metastasis, Lipid Kinases, Metabolic Disorder, Neurobiology, NfkB Pathway, and WNT Signaling research.
PI3K (p85 beta) Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. PI3K (p85 beta) Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~88kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty