Would you like to log in again?
PI3K (p55 gamma) protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S92 009-001-S92S
75930-912EA 665.18 USD InStock
75930-912 75934-094
PI3K (p55 gamma) protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
PI3K (p55 gamma) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PI3K (p55 gamma) also known as PI3K regulatory subunit 3 (PIK3R3) was first identified by 2-hybrid system as a protein that bind to the intracellular domain of IGF1R. PIK3R3 contains 2 Src homology-2 (SH2) domains, an inter-SH2/p110-binding site and a proline-rich motif. PIK3R3 transcript is detected in all fetal and adult tissues except peripheral blood leukocytes. Expression is low in adult liver and most abundant in adult testis. PIK3R3 can bind IGF1R, INSR, and INSR substrate-1 in vitro and the binding requires receptor activation (1). PI3K (p55 gamma) Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Invasion/Metastasis, Lipid Kinases, Metabolic Disorder, Neurobiology, NfkB Pathway, and WNT Signaling research.
PI3K (p55 gamma) Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. PI3K (p55 gamma) Protein is suitable for use in Western Blot. Expect a band approximately ~61kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty