Would you like to log in again?
PCTK1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S83S 009-001-S83
75930-754EA 353.41 USD InStock
75930-754 75930-756
PCTK1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PCTK1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PCTK1 or Pctaire protein kinase 1 belongs to the CDC2/CDKX subfamily of the Ser/Thr family of protein kinases which play a main role in signal transduction cascades in terminally differentiated cells, in exocytosis and in transport of secretory cargo from the endoplasmic reticulum. PCTK1 is ubiquitously expressed with the highest levels detected in the brain and testis (1). PCTK1 maps distal to the t(X;18) synovial sarcoma breakpoint in Xp11.23. A 420-kb YAC clone positive for PCTK1 also contained the gene coding for ubiquitin-activating enzyme UBE1 previously mapped to Xp11.3. (2). PCTK1 is thought to escape X inactivation. PCTK1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
PCTK1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PCTK1 Protein is suitable for use in Western Blot. Expect a band approximately ~62kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty