Would you like to log in again?
PCNA protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S82 009-001-S82S
75930-752 75930-750
PCNA protein-HIS Epitope, Rockland Immunochemicals
PCNA control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PCNA is a nuclear protein whose appearance correlates with the proliferative state of the cells and is a cofactor of DNA polymerase delta. PCNA is a homotrimer and helps increase the processivity of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Immunofluorescence studies have shown that p300 may play a role in DNA repair synthesis through its interaction with PCNA. In vitro and in vivo p300 forms a complex with PCNA that does not depend on the S phase of the cell cycle and stimulates DNA synthesis in vitro (1). PCNA interacts with the Williams syndrome transcription factor (WSTF) allowing it to target to DNA replication foci, that then allows recruitment of SNF2H (2). PCNA Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Cancer, Cell Cycle, ERK/MAPK Pathway, and WNT Signaling research.
PCNA Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. PCNA Protein is suitable for use in Western Blot. Expect a band approximately ~30kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty