Would you like to log in again?
Parkin protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S80 009-001-S80S
75930-738EA 665.18 USD InStock
75930-738 75930-736
Parkin protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
Parkin control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Parkin or parkinson protein 2 is a RING domain-containing E3 ubiquitin protein ligase involved in proteasome-dependent degradation of proteins. Mutations in Parkin can lead to Parkinson’s disease. Parkin is important for mitochondrial quality control by lysosome-dependent degradation of damaged mitochondria through autophagy, or mitophagy (1). Parkin plays an important role in directly supporting mitochondrial function and protecting mitochondrial genomic integrity from oxidative stress (2). Parkin also has essential role in regulating normal respiratory function in mitochondria and in the protection of cells from oxidative stress. PARK7 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Apoptosis/Autophagy, Cancer, Cell Cycle, and Neurobiology research.
Parkin Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. Parkin Protein is suitable for use in Western Blot. Expect a band approximately ~68kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty